3-Bromo-2,6-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A116978-250MG A116978-5G A116978-1G
77189-422EA 29 USD
77189-422 77571-024 77745-863
3-Bromo-2,6-difluorobenzonitrile 97%
3-Bromo-2,6-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
MDL Number: MFCD16987589
CAS Number: 1250444-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR