2-(tert-Butoxy)pyridine-3-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-(tert-Butoxy)pyridin-3-yl)boronic acid, 3-Borono-2-(tert-butoxy)pyridine

A116803-100MG A116803-1G A116803-250MG A116803-5G
77161-054EA 15 USD
77161-054 77161-056 77161-058 77161-060
2-(tert-Butoxy)pyridine-3-boronic acid 97%
2-(tert-Butoxy)pyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₃
MDL Number: MFCD26397235
CAS Number: 1245898-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR