Pyrimidine-4-carbaldehyde oxime 95%

Supplier: Ambeed
Danger

A116570-1G A116570-250MG
77161-034EA 83.43 USD
77161-034 77161-036
Pyrimidine-4-carbaldehyde oxime 95%
Pyrimidine-4-carbaldehyde oxime

Precaución: for Research Use Only

Formula: C₅H₅N₃O
MW: 123.11 g/mol
CAS Number: 1073-65-0
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR