Methyl 3-amino-5-ethoxybenzoate 98%

Supplier: Ambeed
Warning

A116460-1G A116460-250MG A116460-25G A116460-5G
77161-018EA 17.51 USD
77161-018 77161-020 77161-022 77161-024
Methyl 3-amino-5-ethoxybenzoate 98%
Methyl 3-amino-5-ethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.215 g/mol
MDL Number: MFCD20696913
CAS Number: 706792-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR