1,4-Butanesultam 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,2-Thiazinane 1,1-Dioxide
A116368-1G
A116368-250MG
A116368-5G
77189-396EA
34
USD
77189-396
77189-398
77189-400
1,4-Butanesultam 99%
1,4-Butanesultam
Precaución: for Research Use Only
Formula:
C₄H₉NO₂S MW: 135.18 g/mol |
MDL Number:
MFCD04038916 CAS Number: 37441-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...