4-(2-Thienyl)butyric acid 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Thiophen-2-yl)butanoic acid

A116250-10G A116250-1G A116250-5G
77189-384EA 60.77 USD
77189-384 77189-386 77189-388
4-(2-Thienyl)butyric acid 97%
4-(2-Thienyl)butyric acid

Precaución: for Research Use Only

Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
MDL Number: MFCD00005463
CAS Number: 4653-11-6
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR