2-Chloro-5-(trifluoromethyl)benzoic acid 96%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid, 6-Chloro-α,α,α-trifluoro-m-toluic acid

A116121-1G A116121-25G A116121-5G
77189-368EA 22.66 USD
77189-368 77189-370 77189-372 77189-374
2-Chloro-5-(trifluoromethyl)benzoic acid 96%
2-Chloro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD00061094
CAS Number: 657-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR