1,2-Bis(2-chloroethoxy)ethane 98%

Supplier: Ambeed
Danger

A115953-25G A115953-100G
77189-358EA 6.18 USD
77189-358 77614-912
1,2-Bis(2-chloroethoxy)ethane 98%
1,2-Bis(2-chloroethoxy)ethane

Precaución: for Research Use Only

Formula: ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl
MW: 187.07 g/mol
MDL Number: MFCD00000976
CAS Number: 112-26-5
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR