Diethyl cyclohexane-1,1-dicarboxylate 98%

Supplier: Ambeed
Warning

A115438-1G A115438-25G A115438-5G
77189-352EA 7 USD
77189-352 77189-354 77189-356
Diethyl cyclohexane-1,1-dicarboxylate 98%
Diethyl cyclohexane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MDL Number: MFCD02093457
CAS Number: 1139-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR