2-Chloro-1-(3,4-difluorophenyl)ethanol 95%

Supplier: Ambeed
Danger

A115422-100MG A115422-1G A115422-250MG
77160-968EA 67.98 USD
77160-968 77160-970 77160-972
2-Chloro-1-(3,4-difluorophenyl)ethanol 95%
2-Chloro-1-(3,4-difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇ClF₂O
MW: 192.59 g/mol
MDL Number: MFCD14529767
CAS Number: 51336-97-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR