tert-Butyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A115366-100MG A115366-250MG A115366-1G A115366-5G
77160-964EA 37 USD
77160-964 77160-966 77570-904 77570-906
tert-Butyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate 98%
tert-Butyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₈F₃NO₃
MDL Number: MFCD16990768
CAS Number: 550371-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR