Ethyl-2,6-difluorobenzoate 97%

Supplier: Ambeed
Warning

A115261-10G A115261-1G A115261-25G A115261-5G
77160-952EA 12 USD
77160-952 77160-954 77160-956 77160-958
Ethyl-2,6-difluorobenzoate 97%
Ethyl-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MDL Number: MFCD06204338
CAS Number: 19064-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR