Rucaparib 98%

Supplier: Ambeed
Warning

Synonyms: 8-Fluoro-2-(4-((methylamino)methyl)phenyl)-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one

A115210-100MG A115210-10MG A115210-1G A115210-1MG A115210-25MG A115210-5G A115210-5MG A115210-250MG
77160-938EA 35.02 USD
77160-938 77160-940 77160-942 77160-944 77160-946 77160-948 77160-950 77683-049
Rucaparib 98%
Rucaparib

Precaución: For research use only.

Formula: C₁₉H₁₈FN₃O
MW: 323.3641 g/mol
MDL Number: MFCD11977252
CAS Number: 283173-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR