Methyl 5-(benzo[d][1,3]dioxol-5-yl)picolinate 98%
Supplier: Ambeed
|
Warning
|
A114899-100MG
A114899-1G
A114899-250MG
77160-920EA
25
USD
77160-920
77160-922
77160-924
Methyl 5-(benzo[d][1,3]dioxol-5-yl)picolinate 98%
Methyl 5-(benzo[d][1,3]dioxol-5-yl)picolinate
Precaución: for Research Use Only
Formula:
C₁₄H₁₁NO₄ MW: 257.24 g/mol |
MDL Number:
MFCD28902477 CAS Number: 1799412-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...