1,2,3-Butanetriol 90%

Supplier: Ambeed
Warning

Synonyms: 1,2,3-Trihydroxybutane, Butane-1,2,3-triol

A114788-1G A114788-5G
77189-280EA 43 USD
77189-280 77189-282
1,2,3-Butanetriol 90%
1,2,3-Butanetriol

Precaución: for Research Use Only

Formula: C₄H₁₀O₃
MDL Number: MFCD00059662
CAS Number: 4435-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR