Methyl 4-bromo-2-fluoro-3-methylbenzoate 95%
Supplier: Ambeed
|
Warning
|
A114770-1G
A114770-100MG
A114770-250MG
77160-918EA
87
USD
77160-918
77891-822
77891-823
Methyl 4-bromo-2-fluoro-3-methylbenzoate 95%
Methyl 4-bromo-2-fluoro-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrFO₂ |
MDL Number:
MFCD13184289 CAS Number: 1206680-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...