Tropinone 95%

Supplier: Ambeed
Warning

Synonyms: 8-Methyl-8-azabicyclo[3.2.1]octan-3-one

A114731-1000G A114731-100G A114731-10G A114731-25G A114731-500G A114731-5G
77189-268EA 388.31 USD
77189-268 77189-270 77189-272 77189-274 77189-276 77189-278
Tropinone 95%
Tropinone

Precaución: for Research Use Only

Formula: C₈H₁₃NO
MW: 139.2 g/mol
MDL Number: MFCD00005549
CAS Number: 532-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR