4-(4-(Trifluoromethyl)benzyl)piperidine 95%

Supplier: Ambeed
Warning

A114563-1G A114563-250MG
77160-900EA 226.6 USD
77160-900 77683-040
4-(4-(Trifluoromethyl)benzyl)piperidine 95%
4-(4-(Trifluoromethyl)benzyl)piperidine

Precaución: For research use only.

Formula: C₁₃H₁₆F₃N
MW: 243.26 g/mol
MDL Number: MFCD09998992
CAS Number: 192990-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR