1-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A114491-100MG A114491-250MG
77160-896EA 61.8 USD
77160-896 77160-898
1-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid 95%
1-(4-Chlorophenyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O₂
MW: 222.62 g/mol
MDL Number: MFCD10689377
CAS Number: 19532-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR