(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%

Supplier: Ambeed
Danger

A114469-100MG A114469-1G A114469-250MG
77189-262EA 38 USD
77189-262 77189-264 77189-266
(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate ≥98%, ee 99%
(S)-3,3'-Bis(4-nitrophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate

Precaución: for Research Use Only

Formula: C₃₂H₁₉N₂O₈P
MDL Number: MFCD08274951
CAS Number: 878111-16-1
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR