9,9-Dimethylfluorene-2-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 9,9-Dimethylfluorene-2-boronic acid Pinacol ester, 9,9-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A114361-1G A114361-25G A114361-5G
77189-242EA 9.27 USD
77189-242 77189-244 77549-232
9,9-Dimethylfluorene-2-boronic acid pinacol ester 97%
9,9-Dimethylfluorene-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₂₅BO₂
MW: 320.24 g/mol
MDL Number: MFCD08704229
CAS Number: 569343-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR