6-(4-Chlorophenoxy)pyridin-3-amine 97%

Supplier: Ambeed
Warning

A114297-100MG A114297-1G A114297-250MG
77189-236EA 59.74 USD
77189-236 77189-238 77189-240
6-(4-Chlorophenoxy)pyridin-3-amine 97%
6-(4-Chlorophenoxy)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₁₁H₉ClN₂O
MW: 220.655 g/mol
MDL Number: MFCD00052646
CAS Number: 75926-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR