1,8-Octanediol 98%

Supplier: Ambeed
Warning

Synonyms: 1,8-Dihydroxy octane

A114199-100G A114199-10G A114199-25G A114199-500G
77189-220EA 32.96 USD
77189-220 77189-222 77189-224 77566-004
1,8-Octanediol 98%
1,8-Octanediol

Precaución: For research use only.

Formula: C₈H₁₈O₂
MW: 146.23 g/mol
MDL Number: MFCD00002989
CAS Number: 629-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR