2-(Bromomethyl)phenylboronic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)benzeneboronic acid, Dihydroxy(2-(bromomethyl)phenyl)borane, (2-(Bromomethyl)phenyl)metaboric acid, 2-Bromomethylphenylboronic acid

A113992-25G A113992-5G
77189-218EA 131 USD
77189-218 77565-996
2-(Bromomethyl)phenylboronic acid 98%
2-(Bromomethyl)phenylboronic acid

Precaución: For research use only.

Formula: C₇H₈BBrO₂
MDL Number: MFCD01318941
CAS Number: 91983-14-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR