(6-Morpholinopyridazin-3-yl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A113941-100MG A113941-1G
77160-882EA 160 USD
77160-882 77160-884
(6-Morpholinopyridazin-3-yl)methanamine hydrochloride 95%
(6-Morpholinopyridazin-3-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅ClN₄O
MW: 230.70 g/mol
MDL Number: MFCD28134597
CAS Number: 1628557-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR