Ethyl-4,6-dichloro-5-fluoronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,6-dichloro-5-fluoronicotinate

A113860-100MG A113860-1G A113860-250MG A113860-5G
77189-204EA 81.37 USD
77189-204 77189-206 77189-208 77189-210
Ethyl-4,6-dichloro-5-fluoronicotinate 97%
Ethyl-4,6-dichloro-5-fluoronicotinate

Precaución: for Research Use Only

Formula: C₈H₆Cl₂FNO₂
MW: 238.05 g/mol
MDL Number: MFCD18256592
CAS Number: 154012-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR