1,3-Butanediol diacetate 95%

Supplier: Ambeed
Warning

A113759-100G A113759-10G A113759-25G A113759-500G
77189-192EA 57 USD
77189-192 77189-194 77189-196 77189-198
1,3-Butanediol diacetate 95%
1,3-Butanediol diacetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MDL Number: MFCD00008710
CAS Number: 1117-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR