Methyl 2-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline-6-carboxylate 97%

Supplier: Ambeed
Warning

A113659-1G A113659-250MG
77160-876EA 717.91 USD
77160-876 77160-878
Methyl 2-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline-6-carboxylate 97%
Methyl 2-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₃NO₂
MW: 259.22 g/mol
MDL Number: MFCD16251071
CAS Number: 1283718-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR