5-Isopropyl-1H-pyrrole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A113196-1G A113196-250MG
77189-182EA 630.01 USD
77189-182 77189-184
5-Isopropyl-1H-pyrrole-2-carboxylic acid 97%
5-Isopropyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MDL Number: MFCD13174829
CAS Number: 90607-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR