2'-Hydroxy-3'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Hydroxy-3-nitrophenyl)ethanone

A112781-10G A112781-5G A112781-25G A112781-1G
77160-846EA 98.88 USD
77160-846 77160-848 77592-202 77683-032
2'-Hydroxy-3'-nitroacetophenone 98%
2'-Hydroxy-3'-nitroacetophenone

Precaución: For research use only.

Formula: C₈H₇NO₄
MW: 181.1455 g/mol
MDL Number: MFCD02683784
CAS Number: 28177-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR