5-Bromo-3-methoxypyrazin-2(1H)-one 98%

Supplier: Ambeed
Warning

A112630-1G A112630-250MG
77160-820EA 455.01 USD
77160-820 77160-822
5-Bromo-3-methoxypyrazin-2(1H)-one 98%
5-Bromo-3-methoxypyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MDL Number: MFCD22200185
CAS Number: 89282-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR