Empagliflozin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, Jardiance
A112500-10MG
A112500-1G
A112500-250MG
A112500-50MG
A112500-5G
A112500-10G
77160-806EA
5.15
USD
77160-806
77160-808
77160-810
77160-812
77160-814
77570-690
Empagliflozin 98%
Empagliflozin
Precaución: for Research Use Only
Formula:
C₂₃H₂₇ClO₇ MW: 450.9 g/mol |
MDL Number:
MFCD22566222 CAS Number: 864070-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...