2-Amino-4-(4-pyridyl)thiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-4-yl)thiazol-2-amine

A112493-250MG A112493-5G
77160-804EA 18.54 USD
77160-804 77683-028
2-Amino-4-(4-pyridyl)thiazole 97%
2-Amino-4-(4-pyridyl)thiazole

Precaución: For research use only.

Formula: C₈H₇N₃S
MW: 177.22 g/mol
MDL Number: MFCD00490009
CAS Number: 30235-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR