7-Methyl-L-tryptophan 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(7-methyl-1H-indol-3-yl)propanoic acid, H-Trp(7-Me)-OH

A112390-100MG A112390-1MG A112390-250MG
77189-140EA 61.8 USD
77189-140 77189-142 77189-144
7-Methyl-L-tryptophan 97%
7-Methyl-L-tryptophan

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MW: 218.252 g/mol
MDL Number: MFCD06797706
CAS Number: 33468-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR