Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A112319-250MG A112319-1G A112319-5G
77160-802EA 18 USD
77160-802 77570-682 77891-809
Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid 98%
Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD00297221
CAS Number: 378211-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR