(4-Formyl-3,5-dimethoxyphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Formyl-3,5-dimethoxyphenylboronic acid

A112303-100MG A112303-1G A112303-250MG A112303-5G A112303-25G
77160-794EA 13 USD
77160-794 77160-796 77160-798 77160-800 77891-808
(4-Formyl-3,5-dimethoxyphenyl)boronic acid 97%
(4-Formyl-3,5-dimethoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₅
MDL Number: MFCD22383667
CAS Number: 2001080-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR