tert-Butyl 4-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%

Supplier: Ambeed
Warning

A112187-100MG A112187-1G A112187-250MG
77189-128EA 519.12 USD
77189-128 77189-130 77189-132
tert-Butyl 4-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 97%
tert-Butyl 4-chloro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₂O₃
MW: 336.81 g/mol
MDL Number: MFCD27995842
CAS Number: 1707602-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR