Ribavirin 98%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide

A111785-1000G A111785-100G A111785-10G A111785-1MG A111785-25G A111785-500G A111785-5G
77189-090EA 443.93 USD
77189-090 77189-092 77189-094 77189-096 77189-098 77189-100 77189-102
Ribavirin 98%
Ribavirin

Precaución: for Research Use Only

Formula: C8H12N4O5
MW: 244.2047 g/mol
MDL Number: MFCD00058564
CAS Number: 36791-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR