3-Amino-1-(2-methoxyethyl)pyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A111701-1G A111701-250MG
77189-084EA 390.37 USD
77189-084 77189-086
3-Amino-1-(2-methoxyethyl)pyridin-2(1H)-one 97%
3-Amino-1-(2-methoxyethyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
MDL Number: MFCD16679760
CAS Number: 1249057-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR