1,1'-Bis(di-tert-butylphosphino)ferrocene 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF

A111667-10G A111667-1G A111667-250MG A111667-25G A111667-5G A111667-100G
77189-072EA 5.15 USD
77189-072 77189-074 77189-076 77189-078 77189-080 77189-082 77592-192
1,1'-Bis(di-tert-butylphosphino)ferrocene 98%
1,1'-Bis(di-tert-butylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₂₆H₄₄FeP₂
MW: 474.42 g/mol
MDL Number: MFCD01630818
CAS Number: 84680-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR