5-(Aminomethyl)-2-fluorobenzonitrile hydrochloride 97%

Supplier: Ambeed
Warning

A111613-100MG A111613-1G A111613-250MG
77160-772EA 15 USD
77160-772 77160-774 77160-776
5-(Aminomethyl)-2-fluorobenzonitrile hydrochloride 97%
5-(Aminomethyl)-2-fluorobenzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₈H₇FN₂·ClH
MDL Number: MFCD28125524
CAS Number: 1638487-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR