Potassium (N-propylsulfamoyl)amide 99%

Supplier: Ambeed
Danger

A111435-10G A111435-1G A111435-25G A111435-5G
77160-756EA 47.38 USD
77160-756 77160-758 77160-760 77160-762
Potassium (N-propylsulfamoyl)amide 99%
Potassium (N-propylsulfamoyl)amide

Precaución: for Research Use Only

Formula: C₃H₉KN₂O₂S
MW: 176.28 g/mol
MDL Number: MFCD28144694
CAS Number: 1393813-41-6
UN: 2925
ADR: 4.1,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR