(2-(2-Propoxyethoxy)phenyl)boronic acid 95%

Supplier: Ambeed
Danger

A111429-100MG A111429-1G A111429-250MG
77189-064EA 18 USD
77189-064 77189-066 77189-068
(2-(2-Propoxyethoxy)phenyl)boronic acid 95%
(2-(2-Propoxyethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₄
MDL Number: MFCD18434429
CAS Number: 279262-53-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR