5-Bromo-2-chloro-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chloropyridin-3-ol

A111107-100MG A111107-10G A111107-250MG A111107-5G A111107-1G
77160-744EA 7.21 USD
77160-744 77160-746 77160-748 77160-750 77683-017
5-Bromo-2-chloro-3-hydroxypyridine 98%
5-Bromo-2-chloro-3-hydroxypyridine

Precaución: For research use only.

Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
MDL Number: MFCD08688592
CAS Number: 286946-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR