(3-(Ethoxymethyl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A110890-100MG A110890-1G A110890-250MG
77160-724EA 37 USD
77160-724 77160-726 77160-728
(3-(Ethoxymethyl)phenyl)boronic acid 98+%
(3-(Ethoxymethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MDL Number: MFCD08701750
CAS Number: 1107603-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR