5-Methylindole-3-carboxylic acid methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methylindole-3-carboxylate

A110814-100MG A110814-250MG
77160-720EA 27.81 USD
77160-720 77160-722
5-Methylindole-3-carboxylic acid methyl ester 98%
5-Methylindole-3-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
MDL Number: MFCD02179412
CAS Number: 227960-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR