(5-Bromopyrimidin-2-yl)methanamine hydrobromide 97%

Supplier: Ambeed
Warning

A110804-250MG A110804-5G A110804-1G
77160-716EA 97.85 USD
77160-716 77160-718 77570-612
(5-Bromopyrimidin-2-yl)methanamine hydrobromide 97%
(5-Bromopyrimidin-2-yl)methanamine hydrobromide

Precaución: for Research Use Only

Formula: C₅H₇Br₂N₃
MW: 268.93 g/mol
MDL Number: MFCD28144944
CAS Number: 1632285-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR