2-(Methylsulfonyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Methylsulphonyl)benzeneboronic acid, (2-(Methylsulphonyl)phenyl)boronic acid
A110571-100MG
A110571-1G
A110571-250MG
77189-042EA
17.51
USD
77189-042
77189-044
77189-046
2-(Methylsulfonyl)phenylboronic acid 97%
2-(Methylsulphonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₉BO₄S MW: 200.02 g/mol |
MDL Number:
MFCD02179472 CAS Number: 330804-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...