3,5-Bis(trifluoromethyl)chlorobenzene 97%

Supplier: Ambeed
Danger

A110544-100MG A110544-1G A110544-250MG A110544-5G
77189-034EA 12.36 USD
77189-034 77189-036 77189-038 77189-040
3,5-Bis(trifluoromethyl)chlorobenzene 97%
3,5-Bis(trifluoromethyl)chlorobenzene

Precaución: for Research Use Only

Formula: C₈H₃ClF₆
MW: 248.553 g/mol
MDL Number: MFCD01861848
CAS Number: 328-72-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR