5-Ethyl-2-methoxyphenol 97%

Supplier: Ambeed
Warning

A110314-100MG A110314-1G
77160-710EA 70 USD
77160-710 77160-712
5-Ethyl-2-methoxyphenol 97%
5-Ethyl-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD16997738
CAS Number: 2785-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR